missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) chloride, 98%, anhydrous

Product Code. 10114980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10114980 5 g Glass bottle 5g
10308680 100 g Glass bottle 100g
10318680 500 g Glass bottle 500g
13418458 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10114980 Supplier Thermo Scientific Acros Supplier No. 206410050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10026-11-6 | Cl4Zr | 233.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-11-6
Molecular Formula Cl4Zr
Molecular Weight (g/mol) 233.03
MDL Number MFCD00011306
Synonym Zirconium tetrachloride

Specifications

Melting Point 437.0°C
Color White
Physical Form Fine Powder
Quantity 5 g
Packaging Glass bottle
Assay Percent Range 98%
Density 2.8
Linear Formula ZrCl4
Beilstein 01,1295
Fieser 13,16
Merck Index 15,10374
Solubility Information Solubility in water: reacts. Other solubilities: forming hcl and zro2, soluble in alcohol and ether
Formula Weight 233.03
Percent Purity 98%
Chemical Name or Material Zirconium(IV) chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Zirconium(IV) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.