missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) chloride, 98%, cont. 1-2% hafnium(IV) chloride, Thermo Scientific Chemicals

Product Code. 11383716
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383716 250 g 250g
11373716 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383716 Supplier Thermo Scientific Alfa Aesar Supplier No. L14891.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Zirconium(IV) chloride catalyzes the trans-hydrostannylation of acetylenes with Tri-n-butyl­tin hydride. It is used as a superior catalyst for the fries rearrangement of aryl acetates, giving the least hindered ortho-acetyl phenol in high yields at ambient temperatures. In the presence of ZrCl4, chloromethyl ethers are formed under mild conditions from acid chlorides and trioxane or paraformaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium(IV) chloride catalyzes the trans-hydrostannylation of acetylenes with Tri-n-butyl­tin hydride. It is used as a superior catalyst for the fries rearrangement of aryl acetates, giving the least hindered ortho-acetyl phenol in high yields at ambient temperatures. In the presence of ZrCl4, chloromethyl ethers are formed under mild conditions from acid chlorides and trioxane or paraformaldehyde.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-11-6
Molecular Formula ZrCl4
MDL Number MFCD00011306
Synonym Zirconium tetrachloride

Specifications

Melting Point 331°C (sublimation)
Quantity 250 g
UN Number UN2503
Merck Index 14,10174
Solubility Information Reacts with water.
Formula Weight 233.03
Percent Purity 98%
Sensitivity Moisture sensitive
Density 2.803
Assay cont. 1 to 2% hafnium(IV) chloride
Chemical Name or Material Zirconium(IV) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.