missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals

Product Code. 11397237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397237 100 g 100g
11307247 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11397237 Supplier Thermo Scientific Alfa Aesar Supplier No. 011542.22
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
MDL Number MFCD00011307

Specifications

Melting Point 600°C (sublimation)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Insoluble in water.
Formula Weight 167.21
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Hf <0.1%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Chemical Name or Material Zirconium(IV) fluoride, Low hafnium
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.