missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals

Product Code. p-4904582
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11303347

Brand: Thermo Scientific Alfa Aesar 017400.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Light, Air and Moisture Sensitive

Used in the van Arkel-de Boer refining process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-26-0
Molecular Formula I4Zr
MDL Number MFCD00049641
Synonym zirconium iodide, unii-h6nzc1810v, zirconium iv ; iodide, acmc-1bzrf, zirconium 4+ tetraiodide

Specifications

Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99%
Formula Weight 598.84
Chemical Name or Material Zirconium(IV) iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.