missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11420031
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420031 250 g 250g
11410031 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420031 Supplier Thermo Scientific Alfa Aesar Supplier No. 044770.30
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Zirconium(IV) n-propoxide is used as a precursor for the preparation of Zirconia. It is used as coupling agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23519-77-9
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
IUPAC Name propan-1-olate;zirconium(4+)
SMILES CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

Density 1.044 g/mL
Flash Point 28°C (83°F)
Packaging ChemSeal™ bottles
Refractive Index 1.451
Quantity 250 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 327.56
Concentration or Composition (by Analyte or Components) 70% w/w in n-propanol
Physical Form Liquid
Chemical Name or Material Zirconium (IV) n-propoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.