missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 20% in H2O, colloidal dispersion, Thermo Scientific™

Product Code. 11347527
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347527 500 g 500g
11357527 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11347527 Supplier Thermo Scientific Alfa Aesar Supplier No. 012732.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Color White
Physical Form Particles in Liquid
Quantity 500 g
Assay Percent Range 20% in H2O
Solubility Information It is soluble in HF,and hot H2SO4. It is insoluble in water.
Formula Weight 123.22
Odor Recognizable
Density 1.26
Chemical Name or Material colloidal dispersion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.