missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals

Product Code. 10695262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
Product Code. Quantity Packaging unitSize
10695262 250 g Glass bottle 250g
10521451 1 kg Glass bottle 1kg
2 options
This item is not returnable. View return policy

Product Code. 10695262

Brand: Thermo Scientific Acros 190522500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Boiling Point 4300.0°C
Melting Point 2700.0°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98% min. (ZrO2+ HfO2)
Linear Formula ZrO2
Merck Index 15, 10380
Solubility Information Solubility in water: insoluble <1mg/l (20°C). Other solubilities: slightly soluble in hcl and hno3, slowly soluble in hf, disolves on heating with a mixt of 2 parts, h2so4 and 1 part h2o
Formula Weight 123.22
Percent Purity 98.5%
Packaging Glass bottle
Specific Gravity 5.6
Density 5.6000g/mL
Chemical Name or Material Zirconium(IV) oxide, 98.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.