missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 99+% (metals basis excluding Hf), HfO{2} 2%, Thermo Scientific Chemicals

Product Code. 11381189
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Product Code. Quantity unitSize
11381189 250 g 250g
11371189 1 kg 1kg
11391189 5 kg 5kg
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11381189

Brand: Thermo Scientific Alfa Aesar 011395.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium(IV) oxide is used in the production of piezoelectric crystals and also used in the high frequency induction coils and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Boiling Point ca 5000°C
Melting Point ∽2700°C
Physical Form Powder
Quantity 250 g
Assay Percent Range ≥99%, metal basis
Solubility Information Insoluble in water. Soluble in sulfuric acid and hydrofluoric acid.
Formula Weight 123.22
Density 5.89
Chemical Name or Material Zirconium(IV) Oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.