missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals

Product Code. 11389198
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389198 25 g 25g
11399198 1 g 1g
11309208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389198 Supplier Thermo Scientific Alfa Aesar Supplier No. 042583.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2081-12-1
Molecular Formula C16H40O4Zr
Molecular Weight (g/mol) 387.72
MDL Number MFCD00075085
InChI Key WRMYASGYMABHCC-UHFFFAOYSA-N
Synonym zirconium iv tert-butoxide, 2-methylpropan-2-ol; zirconium
PubChem CID 25203936
IUPAC Name 2-methylpropan-2-ol;zirconium
SMILES CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Zr]

Specifications

Color Yellow
Density 0.985 g/mL
Boiling Point 90°C (5 mmHg)
Flash Point 85°C (185°F)
Linear Formula Zr[OC(CH3)3]4
Quantity 25 g
Beilstein 3681870
Sensitivity Light and moisture sensitive
Formula Weight 383.68
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Zirconium (IV) tert-butoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.