missing translation for 'onlineSavingsMsg'
Learn More

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf

Product Code. 10573771 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10573771

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14985-18-3 | N2O7Zr | 231.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14985-18-3
Molecular Formula N2O7Zr
Molecular Weight (g/mol) 231.23
MDL Number MFCD00149896
InChI Key JWBWFTANBHYHBC-UHFFFAOYSA-N
Synonym zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
PubChem CID 16211674
IUPAC Name oxozirconium(2+);dinitrate;hydrate
SMILES [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O

Specifications

Color White
Physical Form Chunks and/or Powder
Quantity 100 g
Assay Percent Range 99.50%
Linear Formula ZrO(NO3)2·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 231.23
Percent Purity 99.5%
Packaging Glass bottle
Chemical Name or Material Zirconyl(IV) nitrate hydrate, 99.5%
compliance-icons compliance-icons
Product Identifier
  • Zirconyl(IV) nitrate hydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.