missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ZLN 024 hydrochloride

Product Code. 18714929
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18714929

Brand: Tocris Bioscience™ 5285/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allosteric AMPK activator

ZLN 024 hydrochloride is an allosteric AMP-activated protein kinase (AMPK) activator; allosterically stimulates active AMPK heterotrimers, increasing the activity of α1β1γ1 by 1.5-fold and α2β1γ1 by 1.7-fold (EC50 values are 0.42 and 0.95 μM, respectively). Inhibits dephosphorylation of AMPK Thr-172 by PP2Cα. Stimulates glucose uptake and fatty acid oxidation without increasing the ADP/ATP ratio. Also decreases fatty acid synthesis and glucose output in primary hepatocytes. Improves glucose tolerance and decreases cholesterol content in db/db mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[2-(2-Bromo-4-methylphenoxy)ethyl]thio]pyrimidine hydrochloride
Quantity 10 mg
Target AMPK Activators
Molecular Formula C13H13BrN2OS.HCl
Formula Weight 361.69
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.