Learn More
Emma Safety Footwear Zolder Safety Shoes
Product Code. 15998641
Change view
Click to view available options
Size:
39
41
43
44
45
46
47
48
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998641 41 1 pair
15988641 39 1 pair
15908651 43 1 pair
15918651 44 1 pair
15928651 45 1 pair
15938651 46 1 pair
15948651 47 1 pair
15958651 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15998641 Supplier Emma Safety Footwear Supplier No. MM50884641
  • ONLINE EXCLUSIVE PRICE
    180.04 €
    104.32 € / 1 pairs
    Save 75.72 €
    42% Off
Estimated Shipment: 18-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Water-repellant oiled Nubuck uppers with internal steel toe protection and an exceptional 'sticky' grip, even on wet and greasy surfaces.
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo/HRO
  • Oiled Nubuck
  • Heat Resistant Outsole
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Brown
Design Zolder
Liner Material Full grain Nappa leather / Split leather
Material Oiled Nubuck
Size 41