missing translation for 'onlineSavingsMsg'
Learn More

Zolmitriptan N-Oxide, TRC

Product Code. 30134538
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134538 25 mg 25mg
17912395 5 mg 5mg
17950805 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134538 Supplier Toronto Research Chemicals Supplier No. TRCZ63901025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251451-30-6
Molecular Formula C16 H21 N3 O3
Molecular Weight (g/mol) 303.36
Synonym 2-Oxazolidinone, 4-[[3-[2-(dimethyloxidoamino)ethyl]-1H-indol-5-yl]methyl]-, (4S)-, (4S)-4-[[3-[2-(Dimethyloxidoamino)ethyl]-1H-indol-5-yl]methyl]-2-oxazolidinone, Zolmitriptan N-Oxide, Imp. B (EP)
IUPAC Name N,N-dimethyl-2-[5-[[(4S)-2-oxo-1,3-oxazolidin-4-yl]methyl]-1H-indol-3-yl]ethanamine oxide
SMILES C[N+](C)([O-])CCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H21N3O3/c1-19(2,21)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-22-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1
Formula Weight 303.1583
Percent Purity >95%
Chemical Name or Material Zolmitriptan N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.