missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide-d4, TRC

Product Code. 30453839
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453839 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30453839 Supplier Toronto Research Chemicals Supplier No. TRCZ7000025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zonisamide-D4
CAS 1020720-04-0
Purity Grade Notes HPLC
Molecular Formula C8 2H4 H4 N2 O3 S
Quantity 5 mg
Synonym Zonisamide-D4
InChI Formula InChI=1S/C8H8N2O3S/c9-14(11,12)5-7-6-3-1-2-4-8(6)13-10-7/h1-4H,5H2,(H2,9,11,12)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c2c(CS(=O)(=O)N)noc2c1[2H]
IUPAC Name (4,5,6,7-tetradeuterio-1,2-benzoxazol-3-yl)methanesulfonamide
Molecular Weight (g/mol) 216.25
Formula Weight 216.0507
Percent Purity >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.