missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide N,N-Dimethylformimidamide, TRC

Product Code. 30009486
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009486 100 mg 100mg
30008827 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009486 Supplier Toronto Research Chemicals Supplier No. TRCZ700015100MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217201-89-2
Molecular Formula C11 H13 N3 O3 S
Molecular Weight (g/mol) 267.3
Synonym N'-(Benzisoxazol-3-ylmethylsulfonyl)-N,N-dimethylformimidamide, Zonisamide N,N-Dimethylformimidamide
IUPAC Name N'-(1,2-benzoxazol-3-ylmethylsulfonyl)-N,N-dimethylmethanimidamide
SMILES CN(C)C=NS(=O)(=O)Cc1noc2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H13N3O3S/c1-14(2)8-12-18(15,16)7-10-9-5-3-4-6-11(9)17-13-10/h3-6,8H,7H2,1-2H3
Formula Weight 267.0678
Chemical Name or Material N'-(Benzisoxazol-3-ylmethylsulfonyl)-N,N-dimethylformimidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.