missing translation for 'onlineSavingsMsg'
Learn More

ZOOM™ Dual Power Supply (220-240 VAC 47 - 60 Hz) œ

Product Code. 10557203
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10557203 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10557203 Supplier Invitrogen Supplier No. ZP10002
  • ONLINE EXCLUSIVE PRICE
    5510.00 €
    4463.10 € / Pack of 1
    Save 1046.90 €
    19% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ZOOM™ Dual Power Supply (220-240 VAC 47 - 60 Hz) œ

The ZOOM™ Dual Power is a microprocessor-controlled programmable power supply capable of running both high voltage-low current and high current-low voltage applications concurrently. The high voltage-low current section is ideal for sample fractionation, IPG strips, IEF, and DNA sequencing applications. The low voltage-high current section is for SDS-PAGE, native PAGE, second-dimension SDS-PAGE, DNA-RNA electrophoresis, and blotting applications. Four sets of output jacks on each side of the unit allow for multiple apparatus to run simultaneously, offering greater workflow efficiency and higher sample throughput. Some of the important features of the ZOOM™ Dual Power unit include:
  • High voltage-low current and low voltage-high current sides can be used independently and concurrently
  • Multiple output jacks allow for the simultaneous running of up to four electrophoresis apparatus on each side of the unit
  • Memory stores up to 30 programs (15 programs per side) of up to six steps per program. Step and ramp functions for discrete and gradual changes of electrical parameters
  • No Load detection alarm system can be disabled to power high voltage applications that require very low current

    Specifications of the ZOOM™ Dual Power
    High voltage-Low current output range




    Voltage: 1–3500 V; adjustable in 1V steps
    Current: 0.01–10.0 mA, adjustable in 0.01 mA increments
    Power: 0.1– 35W in 0.1 W steps

    Low voltage-High current



    output range


    Voltage: 1–250 V; adjustable in 1V steps
    Current: 1–800 mA, adjustable in 1 mA increments
    Power: 1–200 W in 1 W steps

    Power Outlets

    4 pairs of terminals

    Programs Stored

    15 programs, up to 6 steps each

    Duration Limits

    Time: 1 to 9999 min in 1 min steps
    Voltage Hours

    :

    1 to 65000 V- hr in 1 V-hr steps

    Operating Modes

    Constant voltage, constant current, constant power

    Circuit Protection

    Open Circuit; Short Circuit; Thermal Protection Over Voltage, Over Current, Over Power

    Safety

    No Load Detection Enable-Disable Isolated Floating Ground

    Memory Control

    Non-volatile, up to 24 hours. Custom Methods Stored to RAM

    Display

    LCD, view area 111.7 x 86mm

    Temperature

    Ambient 4-40°C

    Dimensions

    23.5 cm (w) x 34.5 cm (h) x 31 cm (d) (37 cm (d) with power cord attached

    Weight

    12 kg (26.5 lbs)

    Applications for the ZOOM™ Dual Power


    High voltage-low current


    Suggested Applications

  • Isoelectric focusing in tube gels
  • Isoelectric focusing in IPG strips
  • Solution phase isoelectric focusing

    Electrophoresis Apparatus:

  • ZOOM™ IPGRunner™ system
  • ZOOM™ IEF Fractionators

    Low voltage-High current
    Suggested Applications

  • Blotting applications
  • DNA and RNA electrophoresis
  • SDS-PAGE
  • Native PAGE

    Electrophoresis Apparatus:

  • XCell SureLock™ Mini-Cell
  • XCell4 SureLock™ Midi-Cell
  • XCell6™ Multigel Unit
  • XCell II™ Blot Module
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance CE, CSA
    Description ZOOM™ Dual Power Supply, 220 to 240V, 47 to 60Hz, LCD Display View area 111.7 x 86mm, For Use With ZOOM™ Dual Power, Used For Protein Assays and Analysis, Protein Gel Electrophoresis
    For Use With (Equipment) Zoom Dual Power
    Timer 1 to 9999 min.
    Type ZOOM™ Dual Power Supply
    Display LCD
    No. of Jacks 4 Sets
    Quantity 1 Ea.
    Temperature (English) Operating 39.2°F to 104°F
    Temperature (Metric) Operating 4°C to 40°C
    Warranty 1 Year
    Width (English) 9.25 in.
    Width (Metric) 23.5 cm
    Height (English) 13.5 in.
    Height (Metric) 34.5 cm
    Weight (English) 26.5 lb.
    Weight (Metric) 12 kg
    Electrical Requirements 220 to 240 V 47 to 60 Hz
    Voltage Input: 220 to 240 V, Output: 1 to 3,500/1 to 250 V
    Wattage 420 w
    Amperage 4 A
    Frequency 47/60 Hz
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.