missing translation for 'onlineSavingsMsg'
Learn More

(±)-Zopiclone-d3 (N-methyl-d3), CDN

Product Code. 30313349
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313349 0.005 g 0.01g
30312931 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30313349 Supplier CDN Supplier No. CDND73850.005G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Tetradecane
CAS 629-59-4
Molecular Formula C14 H30
Quantity 0.005 g
Synonym Tetradecane, Cactus Normal Paraffin N 14, Linpar 14, N 14, NSC 72440, Parafol 14-97, Rubitherm RT 2, TS Paraffin TS 4, n-Tetradecane
InChI Formula InChI=1 S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3
SMILES CCCCCCCCCCCCCC
IUPAC Name tetradecane
Molecular Weight (g/mol) 198.39
Formula Weight 198.2348 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.