Learn More
Tocris Bioscience™ Zosuquidar Trihydrochloride
High affinity P-gp inhibitor
Brand:  Tocris Bioscience™ 5456/50
Encompass
Product Code. 15796846
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Zosuquidar trihydrochloride is a high affinity P-glycoprotein (P-gp) inhibitor (Kd = 79 nM). Restores doxorubicin (Cat. No. 2252) sensitivity in P-gp-expressing multidrug (MDR) resistant cancer cell lines. Also potentiates antitumor efficacy of taxol (Cat. No. 1097) in a MDR human non-small cell lung carcinoma xenograft mouse model.
Specifications
Specifications
Zosuquidar trihydrochloride
(αR)-4-[(1aα,6α,10bα)-1,1-Difluoro-1,1a,6,10b-tetrahydrodibenzo[a,e]cyclopropa[c]cyclohepten-6-yl]-α-[(5-quinolinyloxy)methyl]-1-piperazineethanol trihydrochloride
Soluble to 50mM in DMSO
Bioactive Small Molecule
167465-36-3
50 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Product Title
Select an issue
Research Use Only. Not For Human or Veterinary Use