missing translation for 'onlineSavingsMsg'
Learn More

Zotepine, TRC

Product Code. 30135669
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135669 100 mg 100mg
30009363 250 mg 250mg
30006670 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135669 Supplier Toronto Research Chemicals Supplier No. TRCZ700900100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26615-21-4
Molecular Formula C18 H18 Cl N O S
Molecular Weight (g/mol) 331.86
Synonym 2-[(8-Chlorodibenzo[b,f]thiepin-10-yl)oxy]-N,N-dimethylethanamine, Dibenzo[b,f]thiepin Ethanamine Deriv., Lodopin, Nipolept;
IUPAC Name 2-(3-chlorobenzo[b][1]benzothiepin-5-yl)oxy-N,N-dimethylethanamine
SMILES CN(C)CCOC1=Cc2ccccc2Sc3ccc(Cl)cc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H18ClNOS/c1-20(2)9-10-21-16-11-13-5-3-4-6-17(13)22-18-8-7-14(19)12-15(16)18/h3-8,11-12H,9-10H2,1-2H3
Formula Weight 331.0798
Chemical Name or Material Zotepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.