missing translation for 'onlineSavingsMsg'
Learn More

Zuclopenthixol, TRC

Product Code. 30138316
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138316 10 mg 10mg
17970805 0.5 mg 0.5mg
17983705 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138316 Supplier Toronto Research Chemicals Supplier No. TRCZ70148510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53772-83-1
Molecular Formula C22 H25 Cl N2 O S
Molecular Weight (g/mol) 400.96
Synonym 1-Piperazineethanol, 4-[(3Z)-3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-, 1-Piperazineethanol, 4-[3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-, (Z)-, 4-[(3Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol, 9H-Thioxanthene, 1-piperazineethanol deriv., (Z)-Clopenthixol, Acuphase, Cisordinol, Clopixol, Clopixol depo, Zuclopenthixol, cis-(Z)-Clopenthixol, cis-Clopenthixol, alpha-Clopenthixol
IUPAC Name 2-[4-[(3Z)-3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CC\C=C/2\c3ccccc3Sc4ccc(Cl)cc24)CC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H25ClN2OS/c23-17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)27-22)5-3-9-24-10-12-25(13-11-24)14-15-26/h1-2,4-8,16,26H,3,9-15H2/b18-5-
Formula Weight 400.1376
Chemical Name or Material Zuclopenthixol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.