missing translation for 'onlineSavingsMsg'
Learn More

Zylofuramine, MedChemExpress

Product Code. 16477767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477767 5 mg 5mg
16425048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477767 Supplier Medchem Express Supplier No. HY122477
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Zylofuramine is a psychomotor stimulant.

Zylofuramine is a psychomotor stimulant.

Specifications

Chemical Name or Material Zylofuramine
CAS 3563-92-6
Color White
Purity Grade Notes Research
Molecular Formula C14H21NO
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (455.95 mM)
SMILES CCN[C@@H]([C@@]1([H])CCCO1)CC2=CC=CC=C2
Molecular Weight (g/mol) 219.32
Formula Weight 219.32
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.