missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL
Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Maxymum Recovery, Round
Standard, Round
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 4800
Description
- Tip format: 96 tip and 384 tip
- Tip volume range: 25 μL to 200 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen™ Maxymum Recovery™ surface - Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
- Rack format: Non-ANSI (96) and ANSI/SLAS (384)
- Rack material: Polypropylene
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
| Volume (Metric) | 100 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Rack Color | Gray |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Non-ANSI |
| Format | Rack |
| Tip Style | Maxymum Recovery, Round |
| Packaging | Non-SBS Rack |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction