missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Zymolyase™

Product Code. 11473556 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Concentration:
100,000 U/mg
20,000 U/mg
Product Type:
Zymolyase 100T
Zymolyase 20T
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration Product Type Unit Size
11473556 250 mg 100,000 U/mg Zymolyase 100T 0.25g
12760228 1 g 20,000 U/mg Zymolyase 20T 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473556 Supplier MP Biomedicals™ Supplier No. 08320932
  • ONLINE EXCLUSIVE PRICE
    1505.00 €
    1425.90 € / 0.25g
    Save 79.10 €
    5% Off
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For lysing the cell walls of viable yeast cells and preparing yeast spheroblasts

  • Highly purified preparation from Arthrobacter luteus
  • Efficiently hydrolyzes poly (β-1,3 glucose)-like yeast cell-wall glucan
  • One unit is defined as the amount of activity causing a decrease in A800 of 30%

  • Available with lytic activity of 20,000U/g (Zymolyase 20T) or 100,000U/g (Zymolyase 100T)

Specifications

Concentration 100,000 U/mg
pH 5 to 10
For Use With (Application) Used to lyse living yeast cell walls to produce protoplast or spheroplast of various strains of Yeast cells
Content And Storage 2°C to 8°C
Source Arthrobacter luteus
Quantity 250 mg
Product Type Zymolyase 100T
Form Lyophilized Powder
compliance-icons
Product Identifier
  • Zymolyase 20 T
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Yeast Lytic Enzyme
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.