accessibility menu, dialog, popup

UserName

FFFOHOFFFF

CAS RN 1017777-41-1

IUPAC Name: 2-fluoro-4,6-bis(trifluoromethyl)benzoic acid
Synonyms: 2-fluoro-4,6-bis(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 276.11
Molecular Formula: C9H3F7O2
InChi Key: MRBORTVPNDQHFN-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C(C=C1F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results