accessibility menu, dialog, popup

UserName

H3COClNH2F

CAS RN 1017779-65-5

IUPAC Name: 1-(6-chloro-2-fluoro-3-methoxyphenyl)methanamine
Synonyms: 1-(6-chloro-2-fluoro-3-methoxyphenyl)methanamine
Molecular Weight (g/mol): 189.61
Molecular Formula: C8H9ClFNO
InChi Key: HRDZQFHRDGSFBB-UHFFFAOYSA-N
SMILES: COC1=C(F)C(CN)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results