missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-fluoro-3-methoxybenzylamine, 97%, Thermo Scientific™

Product Code. 15414147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414147 5 g 5g
15404147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414147 Supplier Thermo Scientific Alfa Aesar Supplier No. H31546.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air, carbon dioxide. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-65-5
Molecular Formula C8H9ClFNO
Molecular Weight (g/mol) 189.614
MDL Number MFCD09832379
InChI Key HRDZQFHRDGSFBB-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoro-3-methoxybenzylamine, 6-chloro-2-fluoro-3-methoxyphenyl methanamine, 1-6-chloro-2-fluoro-3-methoxyphenyl methanamine, 6-chloro-2-fluoro-3-methoxybenzylamine, jrd, 6-chloro-2-fluoro-3-methoxyphenyl methylamine, benzenemethanamine, 6-chloro-2-fluoro-3-methoxy
PubChem CID 20112031
IUPAC Name (6-chloro-2-fluoro-3-methoxyphenyl)methanamine
SMILES COC1=C(C(=C(C=C1)Cl)CN)F

Specifications

Melting Point 55°C to 57°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 189.62
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methoxybenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.