accessibility menu, dialog, popup

UserName

H3CONCl

CAS RN 102151-33-7

IUPAC Name: 3-chloro-4-methoxybenzonitrile
Synonyms: 3-chloro-4-methoxybenzonitrile
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: SUFOLDHSHRVSQV-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results