accessibility menu, dialog, popup

UserName

H3COO(E)

CAS RN 103-56-0

IUPAC Name: (2E)-3-phenylprop-2-en-1-yl propanoate
Synonyms: cinnamyl propionate
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: KGDJMNKPBUNHGY-RMKNXTFCSA-N
SMILES: CCC(=O)OC\C=C\C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results