missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl propionate, 97%, Thermo Scientific™

Product Code. 11464637
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464637 100 g 100g
11454637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464637 Supplier Thermo Scientific Alfa Aesar Supplier No. B21142.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-56-0
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00048610
InChI Key KGDJMNKPBUNHGY-RMKNXTFCSA-N
Synonym cinnamyl propionate, cinnamyl alcohol, propionate, propionic acid, cinnamyl ester, 3-phenyl-2-propenyl propionate, 3-phenyl-2-propen-1-yl propionate, 3-phenylallyl propionate, gamma-phenylallyl propionate, 3-phenyl-2-propen-1-ol propanoate, fema no. 2301, 3-phenyl-2-propenyl propanoate
PubChem CID 5355850
IUPAC Name [(E)-3-phenylprop-2-enyl] propanoate
SMILES CCC(=O)OC\C=C\C1=CC=CC=C1

Specifications

Density 1.04
Flash Point 100°C (212°F)
Linear Formula CH3CH2CO2CH2CH=CHC6H5
Quantity 100 g
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material Cinnamyl propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.