accessibility menu, dialog, popup

UserName

CAS RN 103229-89-6

FFFFFFFFOHOF(E/Z)

CAS RN 103229-89-6

IUPAC Name: 2,3,4,5,5,5-hexafluoro-4-(trifluoromethyl)pent-2-enoic acid
Synonyms: 2,3,4,5,5,5-hexafluoro-4-(trifluoromethyl)pent-2-enoic acid
Molecular Weight (g/mol): 276.06
Molecular Formula: C6HF9O2
InChi Key: TWDHAMRDYDLSPH-UHFFFAOYSA-N
SMILES: OC(=O)C(F)=C(F)C(F)(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results