CAS RN 104333-00-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-((2-Nitro-4-(trifluoromethyl)-phenyl)amino)propane-1,2-diol, TRC
CAS: 104333-00-8 Molecular Formula: C10H11N2O4F3 Molecular Weight (g/mol): 280.2 Synonym: 3-[[2-Nitro-4-(trifluoromethyl)phenyl]amino]-1,2-propanediol;3-(4-Trifluoromethyl-2-nitroanilino)propane-1,2-diol,HC Yellow 6,HC Yellow No. 6;N-(2,3-Dihydroxypropyl)-2-nitro-4-(trifluoromethyl)aniline; IUPAC Name: 3-[2-nitro-4-(trifluoromethyl)anilino]propane-1,2-diol SMILES: C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])NCC(CO)O