CAS RN 106881-41-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4,5-Desisopropylidene Topiramate, TRC
CAS: 106881-41-8 Molecular Formula: C9 H17 N O8 S Molecular Weight (g/mol): 299.3 Synonym: 2,3-O-(1-Methylethylidene)-β-D-fructopyranose 1-Sulfamate,5H-1,3-Dioxolo[4,5-b]pyran, β-D-fructopyranose deriv. IUPAC Name: [(3aS,6R,7R,7aS)-6,7-dihydroxy-2,2-dimethyl-5,6,7,7a-tetrahydro-[1,3]dioxolo[4,5-b]pyran-3a-yl]methyl sulfamate SMILES: CC1(C)O[C@H]2[C@H](O)[C@H](O)CO[C@@]2(COS(=O)(=O)N)O1
2
2,3-O-(1-Methylethylidene)-beta-D-fructopyranose 1-Sulfamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.