accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 107-31-3

IUPAC Name: methyl formate
Synonyms: methyl formate
Molecular Weight (g/mol): 60.05
Molecular Formula: C2H4O2
InChi Key: TZIHFWKZFHZASV-UHFFFAOYSA-N
SMILES: COC=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)
  • (3)
  • (1)
Boiling Point 
  • (4)
  • (3)
  • (3)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (10)
  • (1)
  • (3)
  • (1)
  • (1)