missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 97%, pure

Product Code. 10437900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10437900 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10437900 Supplier Thermo Scientific Acros Supplier No. 126820010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-31-3 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Melting Point -100.0°C
Color Colorless
Density 0.9680g/mL
Boiling Point 31.0°C to 33.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HCO2CH3
Refractive Index 1.3420 to 1.3440
Quantity 1 L
Beilstein 02,18
Fieser 05,446
Merck Index 15,6148
Specific Gravity 0.968
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Viscosity 0.35 mPa.s (20°C)
Formula Weight 60.05
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material Methyl formate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl formate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Methyl formate, methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.