accessibility menu, dialog, popup

UserName

CAS RN 107535-73-9

IUPAC Name: 2,3,5,6-tetrafluorobenzoyl chloride
Synonyms: 2,3,5,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol): 212.53
Molecular Formula: C7HClF4O
InChi Key: AELMDUZWKHVPIF-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(C(Cl)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results