accessibility menu, dialog, popup

UserName

CAS RN 109-09-1

IUPAC Name: 2-chloropyridine
Synonyms: 2-chloropyridine C5H4ClN
Molecular Weight (g/mol): 113.54
Molecular Formula: C5H4ClN
InChi Key: OKDGRDCXVWSXDC-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)