accessibility menu, dialog, popup

UserName

BrH3CNCH3CH3H3C

CAS RN 1119-97-7

IUPAC Name: trimethyl(tetradecyl)azanium bromide
Synonyms: myrtrimonium bromide
Molecular Weight (g/mol): 336.4
Molecular Formula: C17H38BrN
InChi Key: CXRFDZFCGOPDTD-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (11)
Quantity 
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (5)
  • (11)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)