accessibility menu, dialog, popup

UserName

CAS RN 112-61-8

IUPAC Name: methyl octadecanoate
Synonyms: methyl stearate
Molecular Weight (g/mol): 298.51
Molecular Formula: C19H38O2
InChi Key: HPEUJPJOZXNMSJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)