missing translation for 'onlineSavingsMsg'
Learn More

Methyl Stearate, TRC

Product Code. 30132534
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132534 10 g 10g
30010972 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132534 Supplier Toronto Research Chemicals Supplier No. TRCM32669010G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-61-8
Molecular Formula C19 H38 O2
Molecular Weight (g/mol) 298.5
Synonym Methyl Stearate, Octadecanoic acid, methyl ester, Stearic acid, methyl ester (6CI,8CI), Emery 2218, Esterol A, Exceparl MS, Kemester 9718, Methyl n-octadecanoate, Methyl octadecanoate, Methyl stearate, NSC 9418, Pastel M 180, Pastell M 180, n-Octadecanoic acid methyl ester
IUPAC Name methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3
Formula Weight 298.2872
Chemical Name or Material Methyl Stearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.