CAS RN 1120-97-4
IUPAC Name:
4-methyl-1,3-dioxane
Synonyms:
4-methyldioxane
Molecular Weight (g/mol):
102.13
Molecular Formula:
C5H10O2
InChi Key:
INCCMBMMWVKEGJ-UHFFFAOYNA-N
SMILES:
CC1CCOCO1
1
–
2
of
2
results
1
Promotions Available
4-Methyl-1,3-dioxane, 99%
4-Methyl-1,3-dioxane, 99%
CAS: 1120-97-4 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 MDL Number: MFCD00006569 InChI Key: INCCMBMMWVKEGJ-UHFFFAOYSA-N Synonym: 4-methyldioxane,4-methyl-m-dioxane,1,3-dioxane, 4-methyl,m-dioxane, 4-methyl,1,3-butanediol formal,4-methyl-1,3 dioxane,acmc-2099cl,5-19-01-00054 beilstein handbook reference,4-methyl-1,3-dioxan-2-yl,m-dioxane, 4-methyl-8ci PubChem CID: 14269 IUPAC Name: 4-methyl-1,3-dioxane SMILES: CC1CCOCO1
2
4-Methyl-1,3-dioxane, TRC
CAS: 1120-97-4 Molecular Formula: C5 H10 O2 Molecular Weight (g/mol): 102.132 Synonym: 4-Methyl-1,3-dioxane (ACI),m-Dioxane, 4-methyl- (6CI, 8CI),4-Methyl-m-dioxane,NSC 292 IUPAC Name: 4-methyl-1,3-dioxane SMILES: CC1CCOCO1