missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-dioxane, 99%

Product Code. 10703541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10703541 50 g Glass bottle 50g
10642793 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703541 Supplier Thermo Scientific Acros Supplier No. 305000500
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1120-97-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-97-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00006569
InChI Key INCCMBMMWVKEGJ-UHFFFAOYSA-N
Synonym 4-methyldioxane, 4-methyl-m-dioxane, 1,3-dioxane, 4-methyl, m-dioxane, 4-methyl, 1,3-butanediol formal, 4-methyl-1,3 dioxane, acmc-2099cl, 5-19-01-00054 beilstein handbook reference, 4-methyl-1,3-dioxan-2-yl, m-dioxane, 4-methyl-8ci
PubChem CID 14269
IUPAC Name 4-methyl-1,3-dioxane
SMILES CC1CCOCO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -45°C to 44°C
Color Colorless
Density 0.9700g/mL
Boiling Point 114°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4140 to 1.416
Quantity 50 g
Specific Gravity 0.97
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1, 3-dioxane, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.