missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-dioxane, TRC

Product Code. 30136652
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136652 5 g 5g
30136398 10 g 10g
30007478 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136652 Supplier Toronto Research Chemicals Supplier No. TRCM3051585G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-97-4
Molecular Formula C5 H10 O2
Molecular Weight (g/mol) 102.132
Synonym 4-Methyl-1,3-dioxane (ACI), m-Dioxane, 4-methyl- (6CI, 8CI), 4-Methyl-m-dioxane, NSC 292
IUPAC Name 4-methyl-1,3-dioxane
SMILES CC1CCOCO1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H10O2/c1-5-2-3-6-4-7-5/h5H,2-4H2,1H3
Formula Weight 102.068
Chemical Name or Material 4-Methyl-1,3-dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.