accessibility menu, dialog, popup

UserName

CAS RN 112282-24-3

NNOClClHCl

CAS RN 112282-24-3

IUPAC Name: 2-(3,4-dichlorophenyl)-1-{2-[(pyrrolidin-1-yl)methyl]piperidin-1-yl}ethan-1-one hydrochloride
Synonyms: 2-(3,4-dichlorophenyl)-1-[2-(pyrrolidin-1-ylmethyl)piperidin-1-yl]ethanone hydrochloride
Molecular Weight (g/mol): 391.76
Molecular Formula: C18H25Cl3N2O
InChi Key: NGVLSOWJSUUYDE-UHFFFAOYNA-N
SMILES: Cl.ClC1=C(Cl)C=C(CC(=O)N2CCCCC2CN2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results