missing translation for 'onlineSavingsMsg'
Learn More

BRL 52537 hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18025672
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025672

Brand: Tocris Bioscience 0699/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective κ agonist

The most κ/μ-selective and among the most potent κ ligands known (25 times more potent than morphine in vivo).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112282-24-3
Molecular Formula C18H25Cl3N2O
Molecular Weight (g/mol) 391.76
MDL Number MFCD00672679
InChI Key NGVLSOWJSUUYDE-UHFFFAOYNA-N
Synonym brl 52537 hydrochloride, 2-3,4-dichlorophenyl-1-2-pyrrolidin-1-ylmethyl piperidin-1-yl ethanone hydrochloride, +/--1-3,4-dichlorophenyl acetyl-2-1-pyrrolidinyl methylpiperidine hydrochloride, 2-3,4-dichlorophenyl-1-2-pyrrolidin-1-ylmethyl piperidin-1-yl ethanone hcl, d03qzj, d04ynw, pleiotropic pathway modulator oral, cancer , celgene, pleiotropic pathway modifiers inflammatory disease , celgene
PubChem CID 11315337
IUPAC Name 2-(3,4-dichlorophenyl)-1-{2-[(pyrrolidin-1-yl)methyl]piperidin-1-yl}ethan-1-one hydrochloride
SMILES Cl.ClC1=C(Cl)C=C(CC(=O)N2CCCCC2CN2CCCC2)C=C1

Specifications

Quantity 50 mg
Formula Weight 391.77
Percent Purity >98%
Chemical Name or Material BRL 52537 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.