CAS RN 1124-39-6
CAS RN 1124-39-6
IUPAC Name:
4-ethylbenzene-1,2-diol
Synonyms:
4-ethylbenzene-1,2-diol
Molecular Weight (g/mol):
138.17
Molecular Formula:
C8H10O2
InChi Key:
HFLGBNBLMBSXEM-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(O)C(O)=C1
1
–
2
of
2
results
1
4-Ethylcatechol, 98%
CAS: 1124-39-6 Molecular Formula: C8H10O2 Molecular Weight (g/mol): 138.17 MDL Number: MFCD00015847 InChI Key: HFLGBNBLMBSXEM-UHFFFAOYSA-N Synonym: 4-ethylcatechol,4-ethylpyrocatechol,4-ethyl-1,2-benzenediol,unii-574jv8byr2,1,2-benzenediol, 4-ethyl,benzenediol, 4-ethyl,3,4-dihydroxyethylbenzene,ethylcatechol,4-ethyl catechol,4-ethylc atechol PubChem CID: 70761 IUPAC Name: 4-ethylbenzene-1,2-diol SMILES: CCC1=CC=C(O)C(O)=C1
2
4-Ethylcatechol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality