accessibility menu, dialog, popup

UserName

OHHOHOO

CAS RN 1143-72-2

IUPAC Name: 4-benzoylbenzene-1,2,3-triol
Synonyms: 4-benzoylbenzene-1,2,3-triol
Molecular Weight (g/mol): 230.22
Molecular Formula: C13H10O4
InChi Key: HTQNYBBTZSBWKL-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results