missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzophenone, 98%, Thermo Scientific™

Product Code. 11433877
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433877 500 g 500g
11413877 25 g 25g
11423877 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433877 Supplier Thermo Scientific Alfa Aesar Supplier No. A18315.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1143-72-2
Molecular Formula C13H10O4
Molecular Weight (g/mol) 230.219
MDL Number MFCD00009996
InChI Key HTQNYBBTZSBWKL-UHFFFAOYSA-N
Synonym 2,3,4-trihydroxybenzophenone, phenyl 2,3,4-trihydroxyphenyl methanone, alizarin yellow a, gallobenzophenone, alizarine yellow a, methanone, phenyl 2,3,4-trihydroxyphenyl, unii-8fbx2q800v, 2,3,4-trihydroxybenzophenon, chembl1527, 2,3,4-trihydroxbenzophenone
PubChem CID 70837
IUPAC Name phenyl-(2,3,4-trihydroxyphenyl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C(=C(C=C2)O)O)O

Specifications

Melting Point 140°C to 142°C
Quantity 500 g
Beilstein 2697065
Formula Weight 230.22
Percent Purity 98%
Chemical Name or Material 2,3,4-Trihydroxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.