accessibility menu, dialog, popup

UserName

CH3CH3OHHC

CAS RN 115-19-5

IUPAC Name: 2-methylbut-3-yn-2-ol
Synonyms: 2-methyl-3-butyn-2-ol
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: CEBKHWWANWSNTI-UHFFFAOYSA-N
SMILES: CC(C)(O)C#C
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)