missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butyn-2-ol, 98%

Product Code. 11371967
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371967 2500 mL 2.5 L
11351967 100 mL 100mL
11361967 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11371967 Supplier Thermo Scientific Alfa Aesar Supplier No. L07593.0F
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 115-19-5 | C5H8O | 84.118 g/mol

2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-19-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00004467
InChI Key CEBKHWWANWSNTI-UHFFFAOYSA-N
Synonym 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol
PubChem CID 8258
IUPAC Name 2-methylbut-3-yn-2-ol
SMILES CC(C)(C#C)O

Specifications

Melting Point 2°C to 3°C
Density 0.863
Boiling Point 103°C to 104°C
Flash Point 22°C (72°F)
Refractive Index 1.421
Quantity 2500 mL
UN Number UN1987
Beilstein 635746
Merck Index 14,6034
Solubility Information Fully miscible.
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 2-Methyl-3-butyn-2-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.