missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butyn-2-ol, 98%

Product Code. 10449260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10449260 1 L Glass bottle 1 L
10502561 250 mL Glass bottle 250mL
11924621 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10449260 Supplier Thermo Scientific Acros Supplier No. 156820010
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 115-19-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-19-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
InChI Key CEBKHWWANWSNTI-UHFFFAOYSA-N
Synonym 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol
PubChem CID 8258
IUPAC Name 2-methylbut-3-yn-2-ol
SMILES CC(C)(C#C)O

Specifications

Melting Point 3.0°C
Color Brown to White
Density 0.8608g/mL
Boiling Point 104.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. tetrahydrofuran (GC)
Packaging Glass bottle
Linear Formula HC≡CC(CH3)2OH
Refractive Index 1.4200 to 1.4220
Quantity 1 L
Beilstein 01,I,235
Fieser 01,674
Merck Index 15,6108
Specific Gravity 0.8608
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,benzene,cellosolve,,carbon tetrachloride,cyclohexanone,diethylene,glycol,ethyl acetate,kerosine,methyl ethyl,ketone,mineral spirits,monoethanolamine
Viscosity 3.02 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2-Methyl-3-butyn-2-ol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-3-butyn-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.